Assay Details
Gene Symbol: HDAC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021069384.1 | XP_020925043.1 | 595 | 60 | ||
XM_021069385.1 | XP_020925044.1 | 578 | 60 | |||
XM_021069386.1 | XP_020925045.1 | 451 | 60 | |||
GenBank mRNA | AK391795.1 | - | 6 - 7 | 542 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
