Assay Details
| Gene Symbol: CDH19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003121695.4 | XP_003121743.1 | 1671 | 97 | ||
| XM_013993286.2 | XP_013848740.1 | 1638 | 97 | |||
| XM_021099400.1 | XP_020955059.1 | 1637 | 97 | |||
| GenBank mRNA | AK391834.1 | - | 6 - 7 | 981 | 97 | |
More Information
Set Membership: |
|

CDH19
Genomic Map