Assay Details
| Gene Symbol: SYNGR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021066554.1 | XP_020922213.1 | 179 | 84 | ||
| GenBank mRNA | AK391847.1 | - | 1 - 2 | 171 | 84 | |
More Information
Set Membership: |
|

SYNGR2
Genomic Map