Assay Details
| Gene Symbol: SETD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003355790.5 | XP_003355838.2 | 949 | 59 | ||
| GenBank mRNA | AK391858.1 | - | 5 - 6 | 1086 | 59 | |
More Information
Set Membership: |
|

SETD6
Genomic Map