Assay Details
| Gene Symbol: ACYP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013992776.2 | XP_013848230.2 | 561 | 76 | ||
| XM_021088228.1 | XP_020943887.1 | 503 | 76 | |||
| XM_021088229.1 | XP_020943888.1 | 491 | 76 | |||
| GenBank mRNA | AK391873.1 | - | 3 - 4 | 293 | 76 | |
More Information
Set Membership: |
|

ACYP2
Genomic Map