Assay Details
| Gene Symbol: LOC100513261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021081221.1 | XP_020936880.1 | 480 | 64 | ||
| XM_021081222.1 | XP_020936881.1 | 247 | 64 | |||
| GenBank mRNA | AK391893.1 | - | 228 | 64 | ||

LOC100513261
Genomic Map