Assay Details
| Gene Symbol: LOC100513261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021081217.1 | XP_020936876.1 | 6920 | 92 | ||
| XM_021081218.1 | XP_020936877.1 | 6994 | 92 | |||
| XM_021081221.1 | XP_020936880.1 | 597 | 92 | |||
| XM_021081222.1 | XP_020936881.1 | 364 | 92 | |||
| GenBank mRNA | AK391893.1 | - | 345 | 92 | ||
More Information
Set Membership: |
|

LOC100513261
Genomic Map