Assay Details
| Gene Symbol: ACTR3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021079295.1 | XP_020934954.1 | 320 | 81 | ||
| XM_021079296.1 | XP_020934955.1 | 320 | 81 | |||
| XM_021079297.1 | XP_020934956.1 | 320 | 81 | |||
| XM_021079298.1 | XP_020934957.1 | 320 | 81 | |||
| XM_021079299.1 | XP_020934958.1 | 321 | 81 | |||
| XM_021079300.1 | XP_020934959.1 | 321 | 81 | |||
| XM_021079301.1 | XP_020934960.1 | 218 | 81 | |||
| XM_021079302.1 | XP_020934961.1 | 320 | 81 | |||
| XM_021079303.1 | XP_020934962.1 | 321 | 81 | |||
| XM_021079304.1 | XP_020934963.1 | 320 | 81 | |||
| XM_021079306.1 | XP_020934965.1 | 320 | 81 | |||
| XM_021079307.1 | XP_020934966.1 | 320 | 81 | |||
| GenBank mRNA | AK391905.1 | - | 3 - 4 | 319 | 81 | |
More Information
Set Membership: |
|

ACTR3B
Genomic Map