Assay Details
| Gene Symbol: B3GNTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003131147.5 | XP_003131195.3 | 794 | 50 | ||
| XM_005656882.3 | XP_005656939.2 | 794 | 50 | |||
| XM_021066333.1 | XP_020921992.1 | 710 | 50 | |||
| XM_021066334.1 | XP_020921993.1 | 710 | 50 | |||
| GenBank mRNA | AK391922.1 | - | 8 - 9 | 760 | 50 | |
More Information
Set Membership: |
|

B3GNTL1
Genomic Map