Assay Details
Gene Symbol: C3H7orf50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013987679.2 | XP_013843133.1 | 275 | 136 | ||
GenBank mRNA | AK391954.1 | - | 1 - 2 | 140 | 136 |

More Information
Set Membership: |
![]() ![]() ![]() |
