Assay Details
| Gene Symbol: SLC39A11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021066701.1 | XP_020922360.1 | 746 | 59 | ||
| XM_021066702.1 | XP_020922361.1 | 979 | 59 | |||
| XM_021066703.1 | XP_020922362.1 | 730 | 59 | |||
| XM_021066704.1 | XP_020922363.1 | 704 | 59 | |||
| XM_021066706.1 | XP_020922365.1 | 2456 | 59 | |||
| XM_021066707.1 | XP_020922366.1 | 587 | 59 | |||
| GenBank mRNA | AK391961.1 | - | 6 - 7 | 717 | 59 | |
More Information
Set Membership: |
|

SLC39A11
Genomic Map