Assay Details
| Gene Symbol: YES1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021096152.1 | XP_020951811.1 | 4221 | 136 | ||
| GenBank mRNA | AK392011.1 | - | 1 | 1224 | 136 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: YES1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021096152.1 | XP_020951811.1 | 4221 | 136 | ||
| GenBank mRNA | AK392011.1 | - | 1 | 1224 | 136 | |
Set Membership: |
|