Assay Details
Gene Symbol: LOC100152306 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005666412.3 | XP_005666469.2 | 66 | |||
XM_021099654.1 | XP_020955313.1 | 66 | ||||
GenBank mRNA | AK392095.1 | - | 1 - 2 | 46 | 66 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005666412.3, XM_021099654.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
