Assay Details
| Gene Symbol: GPAA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003125442.5 | XP_003125490.2 | 1147 | 71 | ||
| GenBank mRNA | AK392160.1 | - | 7 - 8 | 1112 | 71 | |
More Information
Set Membership: |
|

GPAA1
Genomic Map