Assay Details
| Gene Symbol: RAB26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003481110.2 | XP_003481158.1 | 503 | 62 | ||
| GenBank mRNA | AK392200.1 | - | 2 - 3 | 508 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: RAB26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003481110.2 | XP_003481158.1 | 503 | 62 | ||
| GenBank mRNA | AK392200.1 | - | 2 - 3 | 508 | 62 | |
Set Membership: |
|