Assay Details
| Gene Symbol: DBX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021092578.1 | XP_020948237.1 | 974 | 60 | ||
| XM_021092579.1 | XP_020948238.1 | 3511 | 60 | |||
| GenBank mRNA | AK392230.1 | - | 2 - 3 | 541 | 60 | |
More Information
Set Membership: |
|

DBX2
Genomic Map