Assay Details
Gene Symbol: RAB15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021099383.1 | XP_020955042.1 | 844 | 73 | ||
XM_021099384.1 | XP_020955043.1 | 851 | 73 | |||
GenBank mRNA | AK392260.1 | - | 6 - 7 | 544 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
