Assay Details
| Gene Symbol: ADARB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021065068.1 | XP_020920727.1 | 669 | 56 | ||
| XM_021065069.1 | XP_020920728.1 | 669 | 56 | |||
| GenBank mRNA | AK392325.1 | - | 1 - 2 | 205 | 56 | |
More Information
Set Membership: |
|

ADARB2
Genomic Map