Assay Details
Gene Symbol: EMC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003127366.5 | XP_003127414.1 | 525 | 62 | ||
XM_005664785.3 | XP_005664842.1 | 495 | 62 | |||
GenBank mRNA | AK392381.1 | - | 4 - 5 | 439 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
