Assay Details
| Gene Symbol: CYSTM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013995210.2 | XP_013850664.1 | 602 | 96 | ||
| GenBank mRNA | AK392388.1 | - | 2 - 3 | 312 | 96 | |
More Information
Set Membership: |
|

CYSTM1
Genomic Map