Assay Details
| Gene Symbol: NDUFB10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021086798.1 | XP_020942457.1 | 412 | 80 | ||
| GenBank mRNA | AK392464.1 | - | 2 - 3 | 385 | 80 | |
More Information
Set Membership: |
|

NDUFB10
Genomic Map