Assay Details
| Gene Symbol: GNG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013994204.2 | XP_013849658.1 | 499 | 112 | ||
| XM_013994205.2 | XP_013849659.1 | 692 | 112 | |||
| GenBank mRNA | AK392501.1 | - | 1 - 2 | 222 | 112 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013994205.2 transcripts.
Set Membership: |
|

GNG3
Genomic Map