Assay Details
| Gene Symbol: LZTS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005672780.3 | XP_005672837.1 | 1547 | 61 | ||
| XM_005672781.3 | XP_005672838.1 | 1216 | 61 | |||
| XM_013985163.2 | XP_013840617.1 | 4332 | 61 | |||
| GenBank mRNA | AK392511.1 | - | 2 - 3 | 356 | 61 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005672780.3, XM_005672781.3, XM_013985163.2 transcripts.
Set Membership: |
|

LZTS3
Genomic Map