Assay Details
| Gene Symbol: LZTS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005672780.3 | XP_005672837.1 | 2011 | 75 | ||
| XM_005672781.3 | XP_005672838.1 | 1680 | 75 | |||
| XM_005672782.3 | XP_005672839.1 | 1974 | 75 | |||
| XM_013985163.2 | XP_013840617.1 | 4796 | 75 | |||
| XM_021078357.1 | XP_020934016.1 | 2500 | 75 | |||
| GenBank mRNA | AK392511.1 | - | 3 - 4 | 820 | 75 | |
More Information
Set Membership: |
|

LZTS3
Genomic Map