Assay Details
| Gene Symbol: EGFL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005652718.3 | XP_005652775.1 | 362 | 54 | ||
| XM_021081038.1 | XP_020936697.1 | 387 | 54 | |||
| XM_021081039.1 | XP_020936698.1 | 404 | 54 | |||
| XM_021081040.1 | XP_020936699.1 | 144 | 54 | |||
| XM_021081041.1 | XP_020936700.1 | 188 | 54 | |||
| XM_021081042.1 | XP_020936701.1 | 387 | 54 | |||
| XM_021081043.1 | XP_020936702.1 | 502 | 54 | |||
| XM_021081044.1 | XP_020936703.1 | 217 | 54 | |||
| XM_021081045.1 | XP_020936704.1 | 293 | 54 | |||
| XM_021081046.1 | XP_020936705.1 | 179 | 54 | |||
| GenBank mRNA | AK392566.1 | - | 368 | 54 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021081038.1, XM_021081039.1, XM_021081040.1 transcripts.
Set Membership: |
|

EGFL7
Genomic Map