Assay Details
| Gene Symbol: REC114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928745.5 | XP_001928780.1 | 283 | 72 | ||
| XM_005656287.3 | XP_005656344.1 | 285 | 72 | |||
| XM_005656289.3 | XP_005656346.1 | 293 | 72 | |||
| XM_021099081.1 | XP_020954740.1 | 299 | 72 | |||
| GenBank mRNA | AK392585.1 | - | 2 - 3 | 267 | 72 | |
More Information
Set Membership: |
|

REC114
Genomic Map