Assay Details
| Gene Symbol: LOC100739163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021082742.1 | XP_020938401.1 | 163 | 69 | ||
| XM_021082743.1 | XP_020938402.1 | 67 | 69 | |||
| XM_021082744.1 | XP_020938403.1 | 457 | 69 | |||
| GenBank mRNA | AK392586.1 | - | 2 - 3 | 71 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021082743.1 transcripts.
Set Membership: |
|

LOC100739163
Genomic Map