Assay Details
| Gene Symbol: FAM8A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003128189.4 | XP_003128237.2 | 849 | 93 | ||
| XM_021100215.1 | XP_020955874.1 | 852 | 93 | |||
| GenBank mRNA | AK392625.1 | - | 1 - 2 | 829 | 93 | |
More Information
Set Membership: |
|

FAM8A1
Genomic Map