Assay Details
| Gene Symbol: SRP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003130540.6 | XP_003130588.1 | 224 | 79 | ||
| GenBank mRNA | AK392774.1 | - | 1 - 2 | 135 | 79 | |
| AK395687.1 | - | 1 - 2 | 199 | 79 | ||
| AK395905.1 | - | 1 - 2 | 199 | 79 | ||
| AY609488.1 | - | 1 - 2 | 182 | 79 | ||
More Information
Set Membership: |
|

SRP9
Genomic Map