Assay Details
| Gene Symbol: ST3GAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021093794.1 | XP_020949453.1 | 2155 | 69 | ||
| XM_021093795.1 | XP_020949454.1 | 2463 | 69 | |||
| XM_021093796.1 | XP_020949455.1 | 2588 | 69 | |||
| XM_021093797.1 | XP_020949456.1 | 2815 | 69 | |||
| XM_021093798.1 | XP_020949457.1 | 2673 | 69 | |||
| XM_021093799.1 | XP_020949458.1 | 2582 | 69 | |||
| XM_021093800.1 | XP_020949459.1 | 2360 | 69 | |||
| XM_021093801.1 | XP_020949460.1 | 2234 | 69 | |||
| XM_021093802.1 | XP_020949461.1 | 2319 | 69 | |||
| GenBank mRNA | AK392854.1 | - | 4 - 5 | 381 | 69 | |
More Information
Set Membership: |
|

ST3GAL2
Genomic Map