Assay Details
| Gene Symbol: GAPT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021076855.1 | XP_020932514.1 | 4336 | 77 | ||
| XM_021076856.1 | XP_020932515.1 | 4401 | 77 | |||
| XM_021076857.1 | XP_020932516.1 | 4333 | 77 | |||
| XM_021076858.1 | XP_020932517.1 | 4103 | 77 | |||
| GenBank mRNA | AK392859.1 | - | 1 | 1524 | 77 | |
More Information
Set Membership: |
|

GAPT
Genomic Map