Assay Details
| Gene Symbol: CD22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021097121.1 | XP_020952780.1 | 2672 | 69 | ||
| XM_021097123.1 | XP_020952782.1 | 2576 | 69 | |||
| GenBank mRNA | AK392863.1 | - | 13 - 14 | 2571 | 69 | |
More Information
Set Membership: |
|

CD22
Genomic Map