Assay Details
| Gene Symbol: GZF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003134296.6 | XP_003134344.2 | 76 | |||
| GenBank mRNA | AK392882.1 | - | 1 | 620 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: GZF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003134296.6 | XP_003134344.2 | 76 | |||
| GenBank mRNA | AK392882.1 | - | 1 | 620 | 76 | |
Set Membership: |
|