Assay Details
| Gene Symbol: TCP11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013977800.2 | XP_013833254.2 | 1017 | 59 | ||
| XM_013977801.2 | XP_013833255.2 | 762 | 59 | |||
| XM_021098619.1 | XP_020954278.1 | 1545 | 59 | |||
| XM_021098620.1 | XP_020954279.1 | 1360 | 59 | |||
| XM_021098622.1 | XP_020954281.1 | 1400 | 59 | |||
| XM_021098623.1 | XP_020954282.1 | 1298 | 59 | |||
| XM_021098624.1 | XP_020954283.1 | 1305 | 59 | |||
| XM_021098625.1 | XP_020954284.1 | 7071 | 59 | |||
| XM_021098626.1 | XP_020954285.1 | 1200 | 59 | |||
| XM_021098627.1 | XP_020954286.1 | 2021 | 59 | |||
| XM_021098628.1 | XP_020954287.1 | 1121 | 59 | |||
| GenBank mRNA | AK392958.1 | - | 3 - 4 | 401 | 59 | |
More Information
Set Membership: |
|

TCP11
Genomic Map