Assay Details
| Gene Symbol: POLI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021093313.1 | XP_020948972.1 | 928 | 83 | ||
| XM_021093315.1 | XP_020948974.1 | 928 | 83 | |||
| XM_021093318.1 | XP_020948977.1 | 926 | 83 | |||
| XM_021093321.1 | XP_020948980.1 | 933 | 83 | |||
| XM_021093324.1 | XP_020948983.1 | 680 | 83 | |||
| XM_021093327.1 | XP_020948986.1 | 912 | 83 | |||
| XM_021093329.1 | XP_020948988.1 | 798 | 83 | |||
| GenBank mRNA | AK393055.1 | - | 1 - 2 | 65 | 83 | |
More Information
Set Membership: |
|

POLI
Genomic Map