Assay Details
| Gene Symbol: ECPAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021067458.1 | XP_020923117.1 | 1598 | 68 | ||
| XM_021067461.1 | XP_020923120.1 | 1761 | 68 | |||
| XM_021067464.1 | XP_020923123.1 | 1878 | 68 | |||
| XM_021067470.1 | XP_020923129.1 | 1619 | 68 | |||
| XM_021067473.1 | XP_020923132.1 | 1842 | 68 | |||
| GenBank mRNA | AK393275.1 | - | 14 - 15 | 1479 | 68 | |
More Information
Set Membership: |
|

ECPAS
Genomic Map