Assay Details
| Gene Symbol: MBD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021076098.1 | XP_020931757.1 | 4283 | 92 | ||
| XM_021076099.1 | XP_020931758.1 | 4351 | 92 | |||
| XM_021076101.1 | XP_020931760.1 | 3812 | 92 | |||
| XM_021076102.1 | XP_020931761.1 | 17061 | 92 | |||
| XM_021076103.1 | XP_020931762.1 | 4376 | 92 | |||
| XM_021076104.1 | XP_020931763.1 | 4494 | 92 | |||
| XM_021076105.1 | XP_020931764.1 | 4389 | 92 | |||
| XM_021076106.1 | XP_020931765.1 | 3775 | 92 | |||
| XM_021076107.1 | XP_020931766.1 | 4669 | 92 | |||
| XM_021076108.1 | XP_020931767.1 | 4337 | 92 | |||
| XM_021076109.1 | XP_020931768.1 | 4415 | 92 | |||
| XM_021076110.1 | XP_020931769.1 | 4298 | 92 | |||
| XM_021076111.1 | XP_020931770.1 | 3258 | 92 | |||
| XM_021076112.1 | XP_020931771.1 | 4283 | 92 | |||
| XM_021076113.1 | XP_020931772.1 | 4283 | 92 | |||
| XM_021076114.1 | XP_020931773.1 | 4283 | 92 | |||
| XM_021076115.1 | XP_020931774.1 | 4283 | 92 | |||
| GenBank mRNA | AK393332.1 | - | 1 - 2 | 92 | 92 | |
More Information
Set Membership: |
|

MBD5
Genomic Map