Assay Details
| Gene Symbol: GATC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928504.5 | XP_001928539.1 | 528 | 85 | ||
| GenBank mRNA | AK393402.1 | - | 3 - 4 | 376 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: GATC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928504.5 | XP_001928539.1 | 528 | 85 | ||
| GenBank mRNA | AK393402.1 | - | 3 - 4 | 376 | 85 | |
Set Membership: |
|