Assay Details
| Gene Symbol: SHROOM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021080598.1 | XP_020936257.1 | 808 | 76 | ||
| XM_021080599.1 | XP_020936258.1 | 206 | 76 | |||
| XM_021080602.1 | XP_020936261.1 | 838 | 76 | |||
| GenBank mRNA | AK393529.1 | - | 1 - 2 | 153 | 76 | |
More Information
Set Membership: |
|

SHROOM4
Genomic Map