Assay Details
| Gene Symbol: RIMKLA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021096924.1 | XP_020952583.1 | 4194 | 98 | ||
| XM_021096925.1 | XP_020952584.1 | 4212 | 98 | |||
| GenBank mRNA | AK393534.1 | - | 1 | 1611 | 98 | |
More Information
Set Membership: |
|

RIMKLA
Genomic Map