Assay Details
| Gene Symbol: TRMT2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001927785.5 | XP_001927820.3 | 1747 | 63 | ||
| XM_021072787.1 | XP_020928446.1 | 1865 | 63 | |||
| GenBank mRNA | AK393553.1 | - | 9 - 10 | 1723 | 63 | |
More Information
Set Membership: |
|

TRMT2A
Genomic Map