Assay Details
| Gene Symbol: AP2A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021082580.1 | XP_020938239.1 | 2894 | 62 | ||
| XM_021082581.1 | XP_020938240.1 | 2930 | 62 | |||
| XM_021082582.1 | XP_020938241.1 | 7495 | 62 | |||
| GenBank mRNA | AK393723.1 | - | 9 - 10 | 1171 | 62 | |
More Information
Set Membership: |
|

AP2A2
Genomic Map