Assay Details
| Gene Symbol: POLE4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021087319.1 | XP_020942978.1 | 362 | 75 | ||
| XM_021087320.1 | XP_020942979.1 | 360 | 75 | |||
| GenBank mRNA | AK393901.1 | - | 2 - 3 | 340 | 75 | |
More Information
Set Membership: |
|

POLE4
Genomic Map