Assay Details
| Gene Symbol: CHMP4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003128545.5 | XP_003128593.4 | 846 | 105 | ||
| XM_021099213.1 | XP_020954872.1 | 876 | 105 | |||
| GenBank mRNA | AK393910.1 | - | 4 - 5 | 523 | 105 | |
More Information
Set Membership: |
|

CHMP4A
Genomic Map