Assay Details
| Gene Symbol: STBD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003129093.4 | XP_003129141.2 | 1995 | 114 | ||
| GenBank mRNA | AK394077.1 | - | 1 | 524 | 114 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: STBD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003129093.4 | XP_003129141.2 | 1995 | 114 | ||
| GenBank mRNA | AK394077.1 | - | 1 | 524 | 114 | |
Set Membership: |
|