Assay Details
| Gene Symbol: IL22RA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021095535.1 | XP_020951194.1 | 2812 | 87 | ||
| GenBank mRNA | AK394145.1 | - | 1 | 1427 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IL22RA1| Gene Symbol: IL22RA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021095535.1 | XP_020951194.1 | 2812 | 87 | ||
| GenBank mRNA | AK394145.1 | - | 1 | 1427 | 87 | |
Set Membership: |
|