Assay Details
| Gene Symbol: CNOT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001925220.5 | XP_001925255.3 | 1036 | 82 | ||
| GenBank mRNA | AK394153.1 | - | 6 - 7 | 1056 | 82 | |
More Information
Set Membership: |
|

CNOT9
Genomic Map