Assay Details
| Gene Symbol: DISP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003127566.4 | XP_003127614.1 | 3945 | 67 | ||
| XM_005664991.3 | XP_005665048.1 | 3875 | 67 | |||
| GenBank mRNA | AK394365.1 | - | 7 - 8 | 1045 | 67 | |
More Information
Set Membership: |
|

DISP3
Genomic Map