Assay Details
| Gene Symbol: SFPQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021095897.1 | XP_020951556.1 | 1915 | 64 | ||
| XM_021095898.1 | XP_020951557.1 | 1915 | 64 | |||
| GenBank mRNA | AK394449.1 | - | 2 - 3 | 185 | 64 | |
More Information
Set Membership: |
|

SFPQ
Genomic Map