Assay Details
| Gene Symbol: UBQLN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021079831.1 | XP_020935490.1 | 2482 | 85 | ||
| GenBank mRNA | AK394611.1 | - | 1 | 522 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: UBQLN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021079831.1 | XP_020935490.1 | 2482 | 85 | ||
| GenBank mRNA | AK394611.1 | - | 1 | 522 | 85 | |
Set Membership: |
|